Learn More
Metro™ Super Erecta™ Wire Shelf, Metroseal 3 with Microban™ Finish
Catalog No. 19030988
Change view
Click to view available options
Dimensions (L x W):
14 x 30 in.
14 x 36 in.
14 x 60 in.
18 x 24 in.
18 x 30 in.
18 x 36 in.
18 x 54 in.
18 x 60 in.
21 x 42 in.
21 x 54 in.
24 x 30 in.
24 x 36 in.
24 x 42 in.
24 x 48 in.
24 x 54 in.
30 x 48 in.
30 x 60 in.
30 x 72 in.
36 x 60 in.
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
19030988 14 x 30 in.
19030989 14 x 36 in.
19030991 14 x 60 in.
19030993 18 x 24 in.
19030994 18 x 30 in.
19030995 18 x 36 in.
19030996 18 x 54 in.
19030997 18 x 60 in.
19031001 21 x 42 in.
19031003 21 x 54 in.
19031006 24 x 30 in.
19031007 24 x 36 in.
19031008 24 x 42 in.
19031010 24 x 54 in.
19031009 24 x 48 in.
19031012 30 x 60 in.
19031011 30 x 48 in.
19031023 30 x 72 in.
19031025 36 x 60 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Catalog No. 19030988 Supplier Metro™ Supplier No. 1430NK3
Wire shelves are easily adjustable and the Metroseal 3 green epoxy with Microban™ finish promotes easy cleaning. For use with Metro Super Erecta shelving systems.
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Corrosion resistant finish for wet or high humidity environments
  • Antimicrobial is built into finish to keep product clean
Specifications
Product Type Wire Shelf
Dimensions (L x W) 14 x 30 in.
Finish Metroseal 3 with Microban