Learn More
Metro™ Super Erecta™ Wire Shelf, Chrome Finish
Wire shelves in chrome finish allow adjustable configurations when used with Metro Super Erecta shelving systems.
$102.13 - $275.50
Specifications
For Use With (Equipment) Metro™ Super Erecta™ shelving system
Includes Plastic sleeves and wedges
Product Type Wire Shelf
Material Chrome
View More Specs
Products 16
Catalog Number Mfr. No. Dimensions (L x W) Load Bearing Capacity (English) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W) Load Bearing Capacity (English) Price Quantity  
11-926-301
View Documents
Metro™
2460NC
60 x 24 in. (152.4 x 61 cm) 21 lb.
Each for $275.50
Only null left
19-133-862
View Documents
Metro™
1836NC
36 x 18 in. (91.4 x 45.7 cm) 400 lb.
Each for $102.13
Only null left
17-820-347
View Documents
Metro™
3060NC
60 x 30 in. (152.4 x 76 cm) 400 lb.
Each for $102.13
Only null left
11-926-221
View Documents
Metro™
3648NC
48 x 36 in. (121.9 x 91 cm) 600 lb.
Each for $102.13
Only null left
19-133-872
View Documents
Metro™
2436NC
36 x 24 in. (91.4 x 61 cm) 400 lb.
Each for $130.89
Only null left
19-030-954
View Documents
Metro™
1442NC
42 x 14 in. (106.6 x 35.5 cm) 800 lb.
Each for $130.89
Only null left
19-030-955
View Documents
Metro™
1460NC
60 x 14 in. (152.4 x 35.5 cm) 600 lb.
Each for $130.89
Only null left
19-030-957
View Documents
Metro™
2124NC
24 x 21 in. (61 x 53 cm) 800 lb.
Each for $130.89
Only null left
19-030-956
View Documents
Metro™
1824NC
24 x 18 in. (61 x 45.7 cm) 800 lb.
Each for $130.89
Only null left
19-030-961
View Documents
Metro™
2148NC
48 x 21 in. (122 x 53 cm) 800 lb.
Each for $130.89
Only null left
19-030-960
View Documents
Metro™
2142NC
42 x 21 in. (106.6 x 53 cm) 800 lb.
Each for $130.89
Only null left
19-030-963
View Documents
Metro™
2172NC
72 x 21 in. (183 x 53 cm) 600 lb.
Each for $130.89
Only null left
19-030-964
View Documents
Metro™
2430NC
30 x 24 in. (76 x 61 cm) 800 lb.
Each for $130.89
Only null left
19-030-968
View Documents
Metro™
3636NC
36 x 36 in. (91 x 91 cm) 800 lb.
Each for $130.89
Only null left
19-030-967
View Documents
Metro™
3072NC
72 x 30 in. (183 x 76 cm) 600 lb.
Each for $130.89
Only null left
19-030-966
View Documents
Metro™
3036NC
36 x 30 in. (91.4 x 76 cm) 800 lb.
Each for $130.89
Only null left
Description
Description
  • Open-wire design minimizes dust accumulation, allows free circulation of air and greater visibility of stored items
  • Real nickel-chrome finish for dry, low humidity environment
  • Protective lacquer coated
Specifications
Specifications
Metro™ Super Erecta™ shelving system
Plastic sleeves and wedges
Wire Shelf
Chrome
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'