Learn More
Metro™ Super Erecta™ Top Track Stationary Intermediate Unit Kits
High-density, easy access storage
Supplier:  Metro™ TTA21S
 View more versions of this product
Catalog No. 19-138-217
Only null left
Explore available promotions
Includes:
  • 2185mm (86 in.) posts
  • Hardware for connecting stationary intermediate unit to tracks on both sides
Description
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Stationary intermediate unit helps organize Super Erecta, Super Erecta Pro™ and Super Adjustable™ Top-Track storage systems
  • 2185mm (86 in.) high posts
Order Info
Shelves are ordered separately
Specifications
Specifications
Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
21 in.
8 lb.
86 in.
None
Stainless Steel
3.6 kg
Stationary Intermediate Unit Kit
2185 mm
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
53 cm
Product Suggestions
Product Suggestions