Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
High-density, easy access storage.
Supplier: Metro™ TTM21S
Includes:
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
- Mobile units move along anchored track allowing easy access to all shelves
Order Info
- Shelves are ordered separately
- One kit required per mobile unit
Specifications
| Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves | |
| 14.8 kg | |
| Mobile Unit Kit | |
| 59.05 cm | |
| None | |
| Stainless Steel |
| 23.25 in. | |
| 21 in. | |
| 33 lb. | |
| Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system | |
| 53 cm |