Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
High-density, easy access storage.
Supplier: Metro™ TTM24K3
Includes:
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
- Mobile units move along anchored track allowing easy access to all shelves
MetroSeal 3 Finish
Is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
- Shelves are ordered separately
- One kit required per mobile unit
Specifications
| Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves | |
| 15.3 kg | |
| Mobile Unit Kit | |
| 66.7 cm | |
| Metroseal Green Epoxy | |
| Steel |
| 26.25 in. | |
| 24 in. | |
| 34 lb. | |
| Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system | |
| 61 cm |