Learn More
Metro™ Super Erecta™ Top Track Mobile Unit Kits
High-density, easy access storage.
Supplier:  Metro™ TTM21K3
 View more versions of this product
Catalog No. 19-138-223
Only null left
Explore available promotions
Includes:
Posts, casters/caster channels, donut bumpers and roller bearing assemblies
Description
Description
Top-Track storage systems increase storage capacity and ease of access with overhead guide tracks. This design allows for open aisles eliminating hard to reach spaces, an interchangeable/modular storage system and easier cleaning.
  • Mobile units move along anchored track allowing easy access to all shelves
MetroSeal 3 Finish
Is a multi-layer corrosion resistant finish consisting of a proprietary cross linked thermoset epoxy enhanced with Microban antimicrobial protection over a zinc chromate coating. Metroseal 3 with Microban provides protection from bacteria, mold, mildew, and fungi build-up that may cause odor, stains, and product degradation, while displaying excellent abrasion, gouge, and chemical resistance properties.
Order Info
  • Shelves are ordered separately
  • One kit required per mobile unit
Specifications
Specifications
Posts, casters/channel caster, donut bumpers and roller bearing assemblies, hardware, split sleeves
14.8 kg
Mobile Unit Kit
59.05 cm
Metroseal 3 with Microban™
Steel
23.25 in.
21 in.
33 lb.
Metro™ Super Erecta™, Super Erecta Pro™, Super Adjustable™ Top-Track™ storage system
53 cm
Product Suggestions
Product Suggestions