missing translation for 'onlineSavingsMsg'
Learn More

Metro™ Super Erecta™ Solid Shelf, Stainless Steel, Autoclavable and Cart-Washable

Keep autoclave equipment neat and in place. Metro Super Erecta™ Solid Shelf is perfect for autoclave and cart washing applications.

$894.00 - $894.00

Specifications

Product Type Solid Shelf
Color Silver
Material Stainless Steel
Autoclavable Autoclavable
For Use With (Equipment) Metro Super Erecta Shelving System
View More Specs

Products 1
Catalog Number Mfr. No. Width (English) Length (English) Price Quantity  
Catalog Number Mfr. No. Width (English) Length (English) Price Quantity  
13-450-954
View Documents
Metro™
2460NFS
24 in. 60 in. N/A
Only null left
 
Description

Description

  • All-stainless construction
  • Order aluminum split sleeves with stainless C-rings for corrosive, high-temperature autoclave or cart washing applications (13-450-953)
Specifications

Specifications

Solid Shelf
Stainless Steel
Metro Super Erecta Shelving System
Silver
Autoclavable
Videos
SDS
Documents

Documents