Learn More
Metro™ Stainless Lab Worktable, Black Phenolic Top and 3-Sided Frame
Lab worktable helps keep workers safe with a phenolic top and corrosion-proof, stainless steel construction throughout the body.
Supplier:  Metro™ LTS48UPB
 View more versions of this product
Catalog No. 13-451-010
Only null left
Explore available promotions
Description
Description
  • Easily cleaned smooth surfaces
  • Mobile units can be easily moved to clean behind table area
  • Wide range of accessories and caster options available
Order Info
Casters must be ordered separately.
Specifications
Specifications
47.75 in.
47.75 x 30 x 36 in.
91.4 cm
3-sided Frame
273 kg
121.2 cm
Silver
Stationary Table
Laboratory
36 in.
30 in.
76.2 cm
600 lb.
Stainless Steel
Product Suggestions
Product Suggestions