missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MW Series Utility Cart with Wire Shelves
Description
- Cart load capacity 375 lb. (170.1kg)
- Fully interchangeable Super Erecta shelves
- Center shelf can be positioned at any height in 1 in. (2.5 cm) increments
- One-piece stainless-steel handles add rigidity and durability
- 18 in. (46 cm) carts have 4 in. dia. (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. dia. (13 cm) casters
- Three tiers
- Solid Shelves
- 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
- Raised edges help contain spills and make cleanup easier
- Sturdy corner castings facilitate mounting
Order Info
Shipped knocked down. Assembles/disassembles quickly and easily without tools.
Specifications
Specifications
| Product Type | Utility Cart |
| Capacity (Metric) | 170 kg |
| Certifications/Compliance | NSF |
| Color | Silver |
| Description | 3-tier Model |
| Dimensions (L x W x H) | 36 x 24 x 39 in. |
| For Use With (Application) | Transporting Materials |
| Handle Material | Chrome |
| Height (English) | 39 in. |
| Height (Metric) | 99 cm |
| Show More |