missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Cart with Solid and Wire Shelves

Catalog No. 91137
Change view
Click to view available options
Dimensions (L x W x H):
18 x 30 x 39 in.
24 x 18 x 38 in.
36 x 21 x 38 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W x H)
91137 36 x 21 x 38 in.
19123325 18 x 30 x 39 in.
91136 24 x 18 x 38 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 91137 Supplier Metro™ Supplier No. MW404

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Utility cart is a smart addition to any workplace featuring Super Erecta™ shelves with plastic split sleeves, two one-piece handles, and casters with donut bumpers.

  • Stainless steel, chrome shelf and handle material.
  • Load capacity: 375 lb. (170.1 kg).
  • Fully interchangeable Super Erecta™ shelves.
  • Center shelf can be positioned at any height in 1 in. (2.5 cm) increments.
  • One-piece stainless-steel or chrome handles (depends on model) add rigidity and durability.
  • 18 in. (46 cm) carts have 4 in. diameter (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. diameter (13 cm) casters.
  • Three tiers, 375 lbs (170 kg) weight capacity per cart.
  • Solid Shelves 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength.
  • Raised edges help contain spills and make cleanup easier.
  • Sturdy corner castings facilitate mounting.
  • Wire Shelves chrome-plated or stainless-steel shelves with trademarked truss design for added strength.
  • Corners slide easily over special tapered snap-together plastic sleeves to fasten them securely in place.

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Product Type Utility Cart
Capacity (Metric) 170 kg
Certifications/Compliance NSF
Color Silver
Description 3-tier Model
Dimensions (L x W x H) 36 x 21 x 38 in.
For Use With (Application) Transporting Materials
Handle Material Chrome
Height (English) 38 in.
Height (Metric) 97 cm
Includes Super Erecta™ 3 Chrome-plated wire shelves with plastic split sleeves, 2 one-piece Chrome-plated handle, 4 in. dia. caster, 3.5 in. donut bumpers
Length (English) 36 in.
Length (Metric) 91 cm
Load Bearing Capacity (English) 375 lb.
Load Bearing Capacity (Metric) 170 kg
Material Stainless Steel
No. of Shelves 3
Shelf Material Stainless Steel, Chrome
Weight (Metric) 26.8 kg
Weight (English) 59 lb.
Width (English) 21 in.
Width (Metric) 53 cm
Show More Show Less