missing translation for 'onlineSavingsMsg'
Learn More

Metro™ MW Series Utility Carts with Solid Shelves

Catalog No. 91132
Change view
Click to view available options
Height (English):
38 in.
39 in.
Width (English):
18 in.
21 in.
Length (English):
24 in.
36 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Width (English) Length (English)
91132 38 in. 18 in. 24 in.
91133 39 in. 21 in. 36 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 91132 Supplier Metro™ Supplier No. MW203

Utility carts are constructed of Super Erecta™ shelves with plastic split sleeves, two one-piece handles ,and casters with donut bumpers to make mobility of lab equipment easy.

  • Cart load capacity 375 lb. (170.1kg)
  • Fully interchangeable Super Erecta shelves
  • Center shelf can be positioned at any height in 1 in. (2.5 cm) increments
  • One-piece stainless-steel handles add rigidity and durability
  • 18 in. (46 cm) carts have 4 in. dia. (10 cm) casters; 21 in. (53 cm) and 24 in. (61 cm) wide carts have 5 in. dia. (13 cm) casters
  • Three tiers
  • Solid Shelves
  • 18-gauge stainless-steel, doubly thick at the edges for rigidity and strength
  • Raised edges help contain spills and make cleanup easier
  • Sturdy corner castings facilitate mounting

Order Info

Shipped knocked down. Assembles/disassembles quickly and easily without tools.

Specifications

Product Type Utility Cart
Capacity (Metric) 170 kg
Certifications/Compliance NSF
Color Silver
Description 3-tier Model
Dimensions (L x W x H) 24 x 18 x 38 in. (61 x 46 x 97 cm)
For Use With (Application) Transporting materials that require an expanse of flat, solid surface
Height (English) 38 in.
Height (Metric) 97 cm
Includes Super Erecta™ 3 Stainless Steel solid (flat) shelves with plastic split sleeves, 2 one-piece Stainless Steel handles, 4 in. dia. caster, 3.5 in. donut bumpers
Length (English) 24 in.
Length (Metric) 61 cm
Load Bearing Capacity (English) 375 lb.
Load Bearing Capacity (Metric) 170 kg
Material Stainless Steel
No. of Shelves 3
Width (English) 18 in.
Width (Metric) 46 cm
Show More Show Less