missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metro™ MetroMax i Basket Glassware Cart
Level 2 cart with built-in Microban antimicrobial product protection allows arrangement configuration, ensuring storage and movement won't disrupt your workday.
$1,243.96 - $2,076.02
Specifications
| Product Type | Glassware Cart |
|---|---|
| Capacity (Metric) | 362.8 kg |
| Color | Taupe, Blue |
| For Use With (Application) | Glassware Transportation |
| Length (English) | 36 in. |
| Catalog Number | Mfr. No. | Height (English) | Description | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Height (English) | Description | Price | Quantity & Availability | ||||
|
13451092
|
Metro™
GWBX32 |
60 in. | 4-Basket Unit |
Each for $2,076.02
|
|
||||
|
13451091
|
Metro™
GWBX31 |
40 in. | 2-Basket Unit |
Pack of 50 Each for $1,243.96
|
|
||||
Description
- Modular design offers ultimate flexibility.
- Reinforced polymers provide superior strength (up to 800 lb. per shelf).
- Corrosion-proof, suitable for damp and humid conditions.
- Lifetime warranty against rust and corrosion.
- Shelves wipe down easily and have a retaining edge that contains drips and spills.
- Two-basket and four-basket capacity glassware solutions are offered in a spacesaving 18 x 36 in. (457 x 914mm) footprint.
- Solid MetroMax bottom shelf provides spill retention and can be removed from shelf frame for easy cleaning.
- Wide range of accessories available separately, including all-polymer casters for maximum corrosion resistance and premium performance.
Order Info
Casters not included (5PC Series Metro casters are recommended for superior lifetime performance).
Specifications
| Glassware Cart | |
| Taupe, Blue | |
| 36 in. | |
| 800 lb. | |
| Polymer | |
| 45.7 cm |
| 362.8 kg | |
| Glassware Transportation | |
| 91.4 cm | |
| 362.8 kg | |
| 18 in. |