missing translation for 'onlineSavingsMsg'
Learn More

Methylurea, 98%

Catalog No. p-7027767
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2477230 250 g
AAB247720B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2477230 Supplier Thermo Scientific Chemicals Supplier No. B2477230
Only null left

CAS: 598-50-5 | C2H6N2O | 74.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-50-5
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.083
MDL Number MFCD00007950
InChI Key XGEGHDBEHXKFPX-UHFFFAOYSA-N
Synonym 1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
PubChem CID 11719
ChEBI CHEBI:44383
IUPAC Name methylurea
SMILES CNC(=O)N

Specifications

Melting Point 96°C to 101°C
pH 6.7
Odor Odorless
Quantity 250 g
Beilstein 878189
Formula Weight 74.08
Percent Purity 98%
Chemical Name or Material Methylurea
EINECSNumber 209-935-0
RTECSNumber YT7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only