missing translation for 'onlineSavingsMsg'
Learn More

Methylurea, 97%

Catalog No. p-169509
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog No. Quantity Packaging
AC127991000 100 g Plastic bottle
AC127995000 500 g Plastic bottle
2 options

Catalog No. AC127991000

Supplier: Thermo Scientific Chemicals 127991000

Only null left

CAS: 598-50-5 | C2H6N2O | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 598-50-5
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
MDL Number MFCD00007950
InChI Key XGEGHDBEHXKFPX-UHFFFAOYSA-N
Synonym 1-methylurea, n-methylurea, monomethylurea, urea, methyl, methyl urea, urea, n-methyl, methylmocovina, methylmocovina czech, methylharnstoff german, n-methyl urea
PubChem CID 11719
ChEBI CHEBI:44383
IUPAC Name methylurea
SMILES CNC(=O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (HPLC)
Packaging Plastic bottle
Linear Formula CH3NHCONH2
Quantity 100 g
Beilstein 04, 64
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: freely soluble in ethanol
Formula Weight 74.08
Percent Purity 97%
Physical Form Crystalline Solid
Chemical Name or Material Methylurea, 97%
Show More Show Less
EINECSNumber 209-935-0
RTECSNumber YT7175000
TSCA TSCA

RUO – Research Use Only