missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyltriphenylsilane, 97%
CAS: 791-29-7 | C19H18Si | 274.438 g/mol
Supplier: Thermo Scientific Chemicals L1099506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Methyltriphenylsilane | |
| 791-29-7 | |
| C19H18Si | |
| 2940360 | |
| GIGVICQLYWGMGW-UHFFFAOYSA-N | |
| methyl(triphenyl)silane | |
| 522714 | |
| 97% |
| 66°C to 68°C | |
| 5 g | |
| MFCD00041325 | |
| silane, methyltriphenyl, triphenylmethylsilane, methyl triphenyl silane, methyl-tri phenyl silane, methyltriphenylsilane | |
| C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 | |
| 274.438 | |
| 274.44 |
Chemical Identifiers
| 791-29-7 | |
| 274.438 | |
| GIGVICQLYWGMGW-UHFFFAOYSA-N | |
| 522714 | |
| C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3 |
| C19H18Si | |
| MFCD00041325 | |
| silane, methyltriphenyl, triphenylmethylsilane, methyl triphenyl silane, methyl-tri phenyl silane, methyltriphenylsilane | |
| methyl(triphenyl)silane |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only