missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylsilane, 97%

Catalog No. AAL1099514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1099514 25 g
AAL1099503 1 g
AAL1099506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1099514 Supplier Thermo Scientific Chemicals Supplier No. L1099514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 791-29-7 | C19H18Si | 274.438 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 791-29-7
Molecular Formula C19H18Si
Molecular Weight (g/mol) 274.438
MDL Number MFCD00041325
InChI Key GIGVICQLYWGMGW-UHFFFAOYSA-N
Synonym silane, methyltriphenyl, triphenylmethylsilane, methyl triphenyl silane, methyl-tri phenyl silane, methyltriphenylsilane
PubChem CID 522714
IUPAC Name methyl(triphenyl)silane
SMILES C[Si](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 66°C to 68°C
Quantity 25 g
Beilstein 2940360
Formula Weight 274.44
Percent Purity 97%
Chemical Name or Material Methyltriphenylsilane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only