missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyltriethoxysilane, 98%
Description
Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Methyltriethoxysilane is used in production of silicone polymers or silicone resin. It is used in non-metal surface treatment, non-aqueous polymer preparation, manufacture of semi-conductors, coatings and sealants. It finds application as cross linking agent and an adhesion promoter.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
Specifications
Specifications
| Boiling Point | 141°C to 143°C |
| Melting Point | <-50°C |
| Quantity | 250 mL |
| Flash Point | 23°C (73°F) |
| Odor | Mild |
| Refractive Index | 1.3835 |
| Sensitivity | Moisture sensitive |
| Density | 0.897 |
| Linear Formula | CH3Si(OCH2CH3)3 |
| UN Number | UN1993 |
| Show More |
Safety and Handling
| Hazard Category | H226-H315-H319-H335 |
| Hazard Statement | GHS H Statement H226-H335 Flammable liquid and vapor. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 217-983-9 |
| RTECSNumber | VV4640000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only