missing translation for 'onlineSavingsMsg'
Learn More

Methyltriethoxysilane, 98+%

Catalog No. p-169457
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174622500 250 mL Glass bottle
AC174620010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC174622500 Supplier Thermo Scientific Chemicals Supplier No. 174622500
Only null left

CAS: 2031-67-6 | C7H18O3Si | 178.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2031-67-6
Molecular Formula C7H18O3Si
Molecular Weight (g/mol) 178.30
MDL Number MFCD00009066
InChI Key CPUDPFPXCZDNGI-UHFFFAOYSA-N
Synonym methyltriethoxysilane, triethoxy methyl silane, methyl triethoxysilane, silane, triethoxymethyl, triethoxysilylmethane, union carbide a-162, ici-ep 5850, silane, methyltriethoxy, methaneorthosiliconic acid, triethyl ester, mtes
PubChem CID 16241
IUPAC Name triethoxy(methyl)silane
SMILES CCO[Si](C)(OCC)OCC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -40.0°C
Color Colorless
Density 0.8900g/mL
Boiling Point 141.0°C to 143.0°C
Flash Point 23°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.3825 to 1.3845
Linear Formula CH3Si(OC2H5)3
Quantity 250 mL
Beilstein 04,629
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Viscosity 0.6 mPa.s (20°C)
Formula Weight 178.3
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Methyltriethoxysilane, 98+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
EINECSNumber 217-983-9
RTECSNumber VV4640000
TSCA TSCA

RUO – Research Use Only