missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyltri-n-octylammonium Hydrogen Sulfate 97.0+%, TCI America™
$130.48 - $393.32
Chemical Identifiers
| CAS | 59158-14-4 |
|---|---|
| Molecular Formula | C25H55NO4S |
| Molecular Weight (g/mol) | 465.78 |
| MDL Number | MFCD03095545 |
| InChI Key | MWSPFHZPVVWJCO-UHFFFAOYSA-M |
| Synonym | Tri-n-octylmethylammonium Hydrogen Sulfate |
| PubChem CID | 10939560 |
| IUPAC Name | methyltrioctylazanium hydrogen sulfate |
| SMILES | OS([O-])(=O)=O.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC |
Chemical Identifiers
| 59158-14-4 | |
| 465.78 | |
| MWSPFHZPVVWJCO-UHFFFAOYSA-M | |
| 10939560 | |
| OS([O-])(=O)=O.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC |
| C25H55NO4S | |
| MFCD03095545 | |
| Tri-n-octylmethylammonium Hydrogen Sulfate | |
| methyltrioctylazanium hydrogen sulfate |
Specifications
| 59158-14-4 | |
| C25H55NO4S | |
| 1 g | |
| MWSPFHZPVVWJCO-UHFFFAOYSA-M | |
| methyltrioctylazanium hydrogen sulfate | |
| 10939560 | |
| ≥97.0% (N,T) | |
| Methyltri-n-octylammonium Hydrogen Sulfate |
| 65°C | |
| MFCD03095545 | |
| Tri-n-octylmethylammonium Hydrogen Sulfate | |
| OS([O-])(=O)=O.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC | |
| 465.78 | |
| 465.78 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0184
TSCA : No