missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyltri-n-octylammonium Chloride 95.0+%, TCI America™
Supplier: TCI America T1365500G
Specifications
| Methyltri-n-octylammonium Chloride | |
| White-Yellow | |
| MFCD00011862 | |
| 2811 | |
| XKBGEWXEAPTVCK-UHFFFAOYSA-M | |
| methyl(trioctyl)azanium;chloride | |
| 21218 | |
| 404.16 | |
| Crystalline Lumps |
| 5137-55-3 | |
| C25H54ClN | |
| 500 g | |
| methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride | |
| CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.[Cl-] | |
| 404.164 | |
| CHEBI:75286 | |
| ≥95.0% (T) |
Chemical Identifiers
| 5137-55-3 | |
| 404.164 | |
| XKBGEWXEAPTVCK-UHFFFAOYSA-M | |
| 21218 | |
| methyl(trioctyl)azanium;chloride |
| C25H54ClN | |
| MFCD00011862 | |
| methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride | |
| CHEBI:75286 | |
| CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.[Cl-] |
Safety and Handling
EINECSNumber : (2)-0184&(9)-1971
RTECSNumber : BR8575000
TSCA : Yes
Recommended Storage : Refrigerator