missing translation for 'onlineSavingsMsg'
Learn More

Methyltri-n-octylammonium Chloride 95.0+%, TCI America™

Catalog No. T136525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T136525G 25 g
T1365500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T136525G Supplier TCI America Supplier No. T136525G

Chemical Identifiers

CAS 5137-55-3
Molecular Formula C25H54ClN
Molecular Weight (g/mol) 404.164
MDL Number MFCD00011862
InChI Key XKBGEWXEAPTVCK-UHFFFAOYSA-M
Synonym methyltrioctylammonium chloride, aliquat 336, trioctylmethylammonium chloride, methyl trioctyl ammonium chloride, tomac, tricaprylmethylammonium chloride, capriquat, methyltricaprylylammonium chloride, tricaprylylmethylammonium chloride, trioctyl methyl ammonium chloride
PubChem CID 21218
ChEBI CHEBI:75286
IUPAC Name methyl(trioctyl)azanium;chloride
SMILES CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC.[Cl-]

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Formula Weight 404.16
Percent Purity ≥95.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Methyltri-n-octylammonium Chloride
EINECSNumber (2)-0184&(9)-1971
RTECSNumber BR8575000
TSCA Yes
Recommended Storage Refrigerator