missing translation for 'onlineSavingsMsg'
Learn More

Methyltri-n-butylammonium chloride, Aliquat 175 (75% aq. soln.)

Catalog No. AAL1801936
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1801936 500 g
AAL1801909 10 g
AAL1801922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1801936 Supplier Thermo Scientific Chemicals Supplier No. L1801936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 56375-79-2 | C13H30ClN | 235.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 56375-79-2
Molecular Formula C13H30ClN
Molecular Weight (g/mol) 235.84
MDL Number MFCD00011847
InChI Key IPILPUZVTYHGIL-UHFFFAOYSA-M
Synonym tributylmethylammonium chloride, n,n-dibutyl-n-methylbutan-1-aminium chloride, methyltributylammonium chloride, methyl tributyl ammonium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride, unii-az0b34me3u, az0b34me3u, tributyl methyl azanium chloride, 1-butanaminium, n,n-dibutyl-n-methyl-, chloride 1:1, acmc-20aker
PubChem CID 91822
IUPAC Name tributyl(methyl)azanium;chloride
SMILES CCCC[N+](C)(CCCC)CCCC.[Cl-]

Specifications

Melting Point -33°C
Density 0.964
Boiling Point 84°C to 85°C (101 mmHg)
Quantity 500 g
Beilstein 6300212
Sensitivity Hygroscopic
Formula Weight 235.84
Concentration or Composition (by Analyte or Components) 75% aq. soln.
Chemical Name or Material Methyltri-n-butylammonium chloride, Aliquat 175
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 260-135-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only