missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyltri-n-butylammonium chloride, Aliquat 175 (75% aq. soln.)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -33°C |
| Density | 0.964 |
| Boiling Point | 84°C to 85°C (101 mmHg) |
| Quantity | 10 g |
| Beilstein | 6300212 |
| Sensitivity | Hygroscopic |
| Formula Weight | 235.84 |
| Concentration or Composition (by Analyte or Components) | 75% aq. soln. |
| Chemical Name or Material | Methyltri-n-butylammonium chloride, Aliquat 175 |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 260-135-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only