missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylthymol Blue Sodium Salt 95.0+%, TCI America™
$96.94 - $1055.05
Chemical Identifiers
| CAS | 1945-77-3 |
|---|---|
| Molecular Formula | C37H44N2Na4O13S |
| Molecular Weight (g/mol) | 848.779 |
| MDL Number | MFCD00148905 |
| InChI Key | LGVVZVZPOQJZKT-UHFFFAOYSA-N |
| PubChem CID | 131850582 |
| IUPAC Name | 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium |
| SMILES | CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] |
Chemical Identifiers
| 1945-77-3 | |
| 848.779 | |
| LGVVZVZPOQJZKT-UHFFFAOYSA-N | |
| 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium |
| C37H44N2Na4O13S | |
| MFCD00148905 | |
| 131850582 | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] |
Specifications
| 1945-77-3 | |
| MFCD00148905 | |
| LGVVZVZPOQJZKT-UHFFFAOYSA-N | |
| 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium | |
| 131850582 | |
| ≥95.0% (HPLC) | |
| 1 g | |
| Methylthymol Blue Sodium Salt |
| C37H44N2Na4O13S | |
| 3,3′C-Bis[N,N-bis(carboxymethyl)aminomethyl]thymolsulfonphthalein Sodium Salt | |
| CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na] | |
| 848.779 | |
| 844.75 | |
| Black/Green | |
| Crystal/Powder |
Safety and Handling
TSCA : Yes