missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylthymol Blue Sodium Salt 95.0+%, TCI America™
Specifications
Specifications
| Color | Black/Green |
| Synonym | 3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]thymolsulfonphthalein Sodium Salt |
| Quantity | 5 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 844.75 |
| Percent Purity | ≥95.0% (HPLC) |
| Chemical Name or Material | Methylthymol Blue Sodium Salt |
Safety and Handling
| TSCA | Yes |