missing translation for 'onlineSavingsMsg'
Learn More

Methylthymol Blue Sodium Salt 95.0+%, TCI America™

Catalog No. B04781G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B04781G 1 g
B04785G 5 g
B047825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B04781G Supplier TCI America Supplier No. B04781G

Chemical Identifiers

CAS 1945-77-3
Molecular Formula C37H44N2Na4O13S
Molecular Weight (g/mol) 848.779
MDL Number MFCD00148905
InChI Key LGVVZVZPOQJZKT-UHFFFAOYSA-N
PubChem CID 131850582
IUPAC Name 2-[[5-[3-[3-[[bis(carboxymethyl)amino]methyl]-4-hydroxy-2-methyl-5-propan-2-ylphenyl]-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-2-hydroxy-6-methyl-3-propan-2-ylphenyl]methyl-(carboxymethyl)amino]acetic acid;sodium
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)CN(CC(=O)O)CC(=O)O)O)C(C)C)C(C)C)O)CN(CC(=O)O)CC(=O)O.[Na].[Na].[Na].[Na]

Specifications

Synonym 3,3′C-Bis[N,N-bis(carboxymethyl)aminomethyl]thymolsulfonphthalein Sodium Salt
Formula Weight 844.75
Percent Purity ≥95.0% (HPLC)
Color Black/Green
Quantity 1 g
Physical Form Crystal/Powder
Chemical Name or Material Methylthymol Blue Sodium Salt