missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylthymol Blue Sodium Salt, Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Packaging | Glass Bottle |
| Linear Formula | C37H40N2O13SNa4 |
| UN Number | NONH for all modes of transport |
| Quantity | 5 g |
| Formula Weight | 844.74g/mol |
| Chemical Name or Material | Methylthymol Blue Sodium Salt |
Safety and Handling
| EINECSNumber | 217-743-3 |