missing translation for 'onlineSavingsMsg'
Learn More

(Methylthio)acetaldehyde Dimethyl Acetal 98.0+%, TCI America™

Catalog No. p-7133105
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22365G 5 g
M223625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22365G Supplier TCI America Supplier No. M22365G

Chemical Identifiers

CAS 40015-15-4
Molecular Formula C5H12O2S
Molecular Weight (g/mol) 136.209
MDL Number MFCD00009848
InChI Key DVOAQLUDKIFSNB-UHFFFAOYSA-N
PubChem CID 638113
IUPAC Name 1,1-dimethoxy-2-methylsulfanylethane
SMILES COC(CSC)OC

Specifications

Color Yellow
Boiling Point 60°C
Quantity 5 g
UN Number 1993
Synonym methylthio acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylthio ethane, ethane, 1,1-dimethoxy-2-methylthio, 1,1-dimethoxy-2-methylsulfanyl-ethane, ethane,1,1-dimethoxy-2-methylthio, methylmercapto acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylsulfanyl ethane, acmc-209j9v, methylthio acetaldehyde dimethylacetal, 2-methylthio acetaldehyde dimethyl acetal
Formula Weight 136.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (Methylthio)acetaldehyde Dimethyl Acetal