missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfuric acid potassium salt, 97%

Catalog No. p-168673
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414890100 10 g Glass bottle
AC414891000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414890100 Supplier Thermo Scientific Chemicals Supplier No. 414890100
Only null left

CAS: 562-54-9 | CH3KO4S | 150.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 562-54-9
Molecular Formula CH3KO4S
Molecular Weight (g/mol) 150.19
MDL Number MFCD00040472
InChI Key WBGWGHYJIFOATF-UHFFFAOYSA-M
Synonym potassium methyl sulfate, methyl potassium sulfate, methylsulfuric acid potassium salt, unii-9b0jf05o1c, sulfuric acid, monomethyl ester, potassium salt, potassium methyl sulphate, potassium monomethylsulfate, sulfuric acid, monomethyl ester, potassium salt 1:1, 75-93-4 parent, potassium methylsulfate
PubChem CID 2733094
IUPAC Name potassium;methyl sulfate
SMILES [K+].COS([O-])(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 212.0°C to 217.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OSO3K
Quantity 10 g
Fieser 01,1249; 02,446; 04,557; 06,649; 08,354
Merck Index 13, 7731
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 150.19
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methylsulfuric acid potassium salt, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause cancer.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-231-3
TSCA TSCA

RUO – Research Use Only