Learn More
Methylphosphonic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1261906
Description
Methylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethylphosphonic acid is used to produce methyl-phosphonic acid dichloride by chlorination.
Solubility
Soluble in water and alcohol.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
| Methylphosphonic acid | |
| 103°C to 107°C | |
| MFCD00002137 | |
| UN3261 | |
| Hygroscopic | |
| Soluble in water and alcohol. | |
| CP(O)(O)=O | |
| 96.02 | |
| CHEBI:45129 | |
| 98% |
| 993-13-5 | |
| CH5O3P | |
| 5 g | |
| 1739372 | |
| methanephosphonic acid, phosphonic acid, methyl, methyl phosphonic acid, unii-329w4ym10z, phosphonomethyl group, ester group, phosphonomethyl, acmc-20ake9, d02fxw, methylphosphonic acid | |
| YACKEPLHDIMKIO-UHFFFAOYSA-N | |
| methylphosphonic acid | |
| 13818 | |
| 96.03 |
Chemical Identifiers
| 993-13-5 | |
| 96.02 | |
| YACKEPLHDIMKIO-UHFFFAOYSA-N | |
| 13818 | |
| methylphosphonic acid |
| CH5O3P | |
| MFCD00002137 | |
| methanephosphonic acid, phosphonic acid, methyl, methyl phosphonic acid, unii-329w4ym10z, phosphonomethyl group, ester group, phosphonomethyl, acmc-20ake9, d02fxw, methylphosphonic acid | |
| CHEBI:45129 | |
| CP(O)(O)=O |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 213-607-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only