missing translation for 'onlineSavingsMsg'
Learn More

Methylmalonic Acid 98.0+%, TCI America™

Supplier:  TCI America M0568100G

 View more versions of this product

Catalog No. M0568100G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Methylmalonic Acid
135°C
C4H6O4
100 g
ZIYVHBGGAOATLY-UHFFFAOYSA-N
2-methylpropanedioic acid
487
118.09
Crystalline Powder
516-05-2
White
MFCD00002656
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
CC(C(O)=O)C(O)=O
118.09
CHEBI:30860
≥98.0% (T)

Chemical Identifiers

516-05-2
118.09
ZIYVHBGGAOATLY-UHFFFAOYSA-N
487
2-methylpropanedioic acid
C4H6O4
MFCD00002656
methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
CHEBI:30860
CC(C(O)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : OO1400000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions