missing translation for 'onlineSavingsMsg'
Learn More

Methylmalonic Acid 98.0+%, TCI America™

Catalog No. M056825G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M056825G 25 g
M0568100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M056825G Supplier TCI America Supplier No. M056825G

Chemical Identifiers

CAS 516-05-2
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002656
InChI Key ZIYVHBGGAOATLY-UHFFFAOYSA-N
Synonym methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
PubChem CID 487
ChEBI CHEBI:30860
IUPAC Name 2-methylpropanedioic acid
SMILES CC(C(O)=O)C(O)=O

Specifications

Melting Point 135°C
Color White
Quantity 25 g
Formula Weight 118.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylmalonic Acid
RTECSNumber OO1400000
TSCA No