missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™

Catalog No. AC427408000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427408000 800 mL AcroSeal™ Glass Bottle
AC427401000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427408000 Supplier Thermo Scientific Chemicals Supplier No. 427408000

CAS: 917-64-6 | CH3IMg | 166.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 917-64-6 , 60-29-7
Molecular Formula CH3IMg
Molecular Weight (g/mol) 166.24
MDL Number MFCD00001026
InChI Key AUPXBVDHVRZMIB-UHFFFAOYSA-M
Synonym methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide
PubChem CID 101907
IUPAC Name magnesium;carbanide;iodide
SMILES C[Mg]I

Specifications

Density 1.2600g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3MgI
Quantity 800 mL
Specific Gravity 1.26
Formula Weight 166.24
Chemical Name or Material Methylmagnesium iodide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed. Causes severe skin burns and eye damage. May cause drowsiness or dizziness. In contact with water releases flammable gases which may ignite spontaneously. Extremely flammable liquid and vapor. May form explosive peroxides. Reacts violently with water. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
Wear eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed.
EINECSNumber 213-031-1

RUO – Research Use Only