missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium bromide, 3.2M solution in 2-MeTHF, AcroSeal™

Catalog No. p-4258690
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC391121000 100 mL AcroSeal™ Glass bottle
AC391128000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC391121000 Supplier Thermo Scientific Chemicals Supplier No. 391121000

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75-16-1 , 96-47-9
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide, grignard reagent
PubChem CID 6349
SMILES C[Mg]Br

Specifications

Color Brown
Physical Form Liquid
Quantity 100 mL
Linear Formula CH3MgBr
Flash Point −11°C
IUPAC Name bromo(methyl)magnesium
Formula Weight 119.24
Concentration or Composition (by Analyte or Components) 34 to 36%
Packaging AcroSeal™ Glass bottle
Chemical Name or Material Methylmagnesium bromide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
Harmful if swallowed.
Reacts violently with water.<b
Precautionary Statement GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 200-844-1

RUO – Research Use Only