missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylmagnesium bromide, 3.2M solution in 2-MeTHF, AcroSeal™
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Brown |
| Physical Form | Liquid |
| Quantity | 800 mL |
| Flash Point | −11°C |
| Packaging | AcroSeal™ Glass Bottle |
| Linear Formula | CH3MgBr |
| IUPAC Name | magnesium;carbanide;bromide |
| Formula Weight | 119.24 |
| Concentration or Composition (by Analyte or Components) | 34 to 36% |
| Chemical Name or Material | Methylmagnesium bromide |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. In contact with water releases flammable gases which may ignite spontaneously. Highly flammable liquid and vapor. Harmful if swallowed. Reacts violently with water.<b |
| Precautionary Statement | GHS P Statement IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for |
| EINECSNumber | 200-844-1 |
RUO – Research Use Only