missing translation for 'onlineSavingsMsg'
Learn More

Methylionone (mixture of alpha- and beta-, predominantly alpha-n-isomer) 80.0+%, TCI America™

Catalog No. M036325ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M036325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M036325ML Supplier TCI America Supplier No. M036325ML
Only null left

Chemical Identifiers

CAS 7779-30-8
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.329
MDL Number MFCD00031478
InChI Key VPKMGDRERYMTJX-CMDGGOBGSA-N
PubChem CID 5371084
IUPAC Name (E)-1-(2,6,6-trimethylcyclohex-2-en-1-yl)pent-1-en-3-one
SMILES CCC(=O)C=CC1C(=CCCC1(C)C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 206.33
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylionone (mixture of alpha- and beta-, predominantly alpha-n-isomer)
EINECSNumber (3)-3305&(3)-3306
TSCA Yes