missing translation for 'onlineSavingsMsg'
Learn More

Methylglyoxal, MP Biomedicals™

Catalog No. p-4405130
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15555825 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15555825 Supplier MP Biomedicals Inc Supplier No. 0215555825

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 78-98-8
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00006960
InChI Key AIJULSRZWUXGPQ-UHFFFAOYSA-N
Synonym methylglyoxal, pyruvaldehyde, pyruvic aldehyde, acetylformaldehyde, acetylformyl, propanal, 2-oxo, 2-ketopropionaldehyde, propanedione, propanolone, methyl glyoxal
PubChem CID 880
ChEBI CHEBI:17158
IUPAC Name 2-oxopropanal
SMILES CC(=O)C=O

Specifications

Color Colorless
Density 1.17g/mL
Quantity 25 mL
Formula Weight 72.06
Physical Form Liquid
Chemical Name or Material Methylglyoxal, Molecular biology
EINECSNumber 201-164-8
Recommended Storage Store at 4°C, dessicated.