Learn More
Methyl thioglycolate, 98%
CAS: 2365-48-2 | C3H6O2S | 106.14 g/mol
$60.20 - $180.40
Chemical Identifiers
| CAS | 2365-48-2 |
|---|---|
| Molecular Formula | C3H6O2S |
| Molecular Weight (g/mol) | 106.14 |
| MDL Number | MFCD00004873 |
| InChI Key | MKIJJIMOAABWGF-UHFFFAOYSA-N |
| Synonym | methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el |
| PubChem CID | 16907 |
| IUPAC Name | methyl 2-sulfanylacetate |
| SMILES | COC(=O)CS |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC148881000
|
Thermo Scientific Chemicals
148881000 |
100 g | Glass bottle |
Each for $60.20
|
|
||||
|
AC148885000
|
Thermo Scientific Chemicals
148885000 |
500 g | Glass bottle |
Each for $180.40
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2365-48-2 | |
| 106.14 | |
| MKIJJIMOAABWGF-UHFFFAOYSA-N | |
| 16907 | |
| COC(=O)CS |
| C3H6O2S | |
| MFCD00004873 | |
| methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el | |
| methyl 2-sulfanylacetate |
Specifications
| 2365-48-2 | |
| Colorless | |
| 156°C to 159°C (10.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| 1.4640 to 1.467 | |
| MFCD00004873 | |
| 1.187 | |
| Solubility in water: 40g/l water (20°C) | |
| COC(=O)CS | |
| 106.14 | |
| 106.14 | |
| Liquid |
| -24°C | |
| 1.1870g/mL | |
| 56°C | |
| 94% min. (Iodimetry) | |
| C3H6O2S | |
| HSCH2CO2CH3 | |
| 100 g | |
| methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el | |
| MKIJJIMOAABWGF-UHFFFAOYSA-N | |
| methyl 2-sulfanylacetate | |
| 16907 | |
| 95% | |
| Methyl Thioglycolate |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 219-121-7
RUO – Research Use Only