missing translation for 'onlineSavingsMsg'
Learn More

Methylenediphosphonic Acid 97.0+%, TCI America™

Catalog No. p-7132397
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08431G 1 g
M08435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08431G Supplier TCI America Supplier No. M08431G
Only null left

Chemical Identifiers

CAS 1984-15-2
Molecular Formula CH4Na2O6P2
Molecular Weight (g/mol) 219.96
MDL Number MFCD00002139
InChI Key RXSYPQBNTPQOTQ-UHFFFAOYSA-L
Synonym methylenediphosphonic acid, medronic acid, acido medronico, methylene diphosphonate, acide medronique, acidum medronicum, phosphonic acid, methylenebis, methylenebis phosphonic acid, mdp-bracco, methanediphosphonic acid
PubChem CID 16124
ChEBI CHEBI:43945
IUPAC Name disodium hydrogen (hydrogen phosphonatomethyl)phosphonate
SMILES [Na+].[Na+].OP([O-])(=O)CP(O)([O-])=O

Specifications

Melting Point 204°C
Quantity 1 g
UN Number 3261
Formula Weight 176.00
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylenediphosphonic Acid
RTECSNumber SZ9138000
TSCA No
Recommended Storage Refrigerator