missing translation for 'onlineSavingsMsg'
Learn More

Methylenediphosphonic acid, 98+%

Catalog No. AAA1480306
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1480306 5 g
AAA1480314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1480306 Supplier Thermo Scientific Chemicals Supplier No. A1480306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Methylenediphosphonic acid, >-99%, CH6O6P2, CAS Number-1984-15-2, acido medronico, methylenediphosphonic acid, acide medronique, acidum medronicum, phosphonic acid, methylenebis, methylenebis phosphonic acid, methylene diphosphonate, mdp-bracco, methanediphosphonic acid, medronic acid, 25g, 1708494 | CAS: 1984-15-2 | CH4Na2O6P2 | 219.96 g/mol

Methylenediphosphonic acid is used as a radioactive imaging agent, which is primarily used in skeletal scintigraphy. Due to its absorption by a variety of tumors, it is useful for the detection of neoplasms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylenediphosphonic acid is used as a radioactive imaging agent, which is primarily used in skeletal scintigraphy. Due to its absorption by a variety of tumors, it is useful for the detection of neoplasms.

Solubility
Soluble in water, dimethylsulfoxide and methanol. Slightly soluble in anhydrous ethanol. Insoluble in methylene chloride.

Notes
Store in a cool place. Incompatible with strong bases.

Chemical Identifiers

CAS 1984-15-2
Molecular Formula CH6O6P2
Molecular Weight (g/mol) 176.00
MDL Number MFCD00002139
InChI Key MBKDYNNUVRNNRF-UHFFFAOYSA-N
Synonym methylenediphosphonic acid, medronic acid, acido medronico, methylene diphosphonate, acide medronique, acidum medronicum, phosphonic acid, methylenebis, methylenebis phosphonic acid, mdp-bracco, methanediphosphonic acid
PubChem CID 16124
ChEBI CHEBI:43945
IUPAC Name (phosphonomethyl)phosphonic acid
SMILES OP(O)(=O)CP(O)(O)=O

Specifications

Melting Point 202°C to 204°C
Odor Odorless
Quantity 5 g
UN Number UN3261
Beilstein 1708494
Sensitivity Hygroscopic
Merck Index 14,5792
Solubility Information Soluble in water,dimethylsulfoxide and methanol. Slightly soluble in anhydrous ethanol. Insoluble in methylene chloride.
Formula Weight 176
Percent Purity ≥99%
Chemical Name or Material Methylenediphosphonic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 217-851-0
RTECSNumber SZ9138000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only