missing translation for 'onlineSavingsMsg'
Learn More

Methylenecyclopentane 98.0+%, TCI America™

Catalog No. p-7132440
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09141ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M09141ML Supplier TCI America Supplier No. M09141ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1528-30-9
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00001421
InChI Key NFJPEKRRHIYYES-UHFFFAOYSA-N
PubChem CID 73714
IUPAC Name methylidenecyclopentane
SMILES C=C1CCCC1

Specifications

Color Colorless
Boiling Point 77°C
Quantity 1 mL
UN Number 3295
Formula Weight 82.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Methylenecyclopentane
TSCA Yes
Recommended Storage Refrigerator