missing translation for 'onlineSavingsMsg'
Learn More

Methylenecyclohexane, 98%

Catalog No. p-3758396
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC325510010 1 g Glass bottle
AC325510050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC325510010 Supplier Thermo Scientific Chemicals Supplier No. 325510010
Only null left

CAS: 1192-37-6 | C7H12 | 96.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1192-37-6
Molecular Formula C7H12
Molecular Weight (g/mol) 96.17
MDL Number MFCD00001661
InChI Key YULMNMJFAZWLLN-UHFFFAOYSA-N
Synonym methylenecyclohexane, cyclohexane, methylene, 1-methylenecyclohexane, methylene-cyclohexane, cyclohexylmethyl, methylcyclohexyl, acmc-209a1w, methylene cyclohexane
PubChem CID 14502
IUPAC Name methylidenecyclohexane
SMILES C=C1CCCCC1

Specifications

Color Colorless
Density 0.8000g/mL
Boiling Point 102.0°C to 103.0°C
Flash Point −006°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4465 to 1.4505
Linear Formula C6H10(=CH2)
Quantity 1 g
Beilstein 05, 69
Specific Gravity 0.8
Formula Weight 96.17
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methylenecyclohexane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-752-4

RUO – Research Use Only