missing translation for 'onlineSavingsMsg'
Learn More

(Methyleneamino)acetonitrile, TCI America™

Catalog No. M021225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M021225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M021225G Supplier TCI America Supplier No. M021225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-82-0
Molecular Formula C3H4N2
Molecular Weight (g/mol) 68.079
MDL Number MFCD00001889
InChI Key GFZMFCVDDFHSJK-UHFFFAOYSA-N
Synonym methyleneaminoacetonitrile, methylenaminoacetonitrile, maan, acetonitrile, methyleneamino, methyleniminoacetonitrile, glycinonitrile, n-methylene, n-methyleneglycinonitrile, 2-methylideneamino acetonitrile, methylenamino acetonitrile, usaf do-5
PubChem CID 8015
IUPAC Name 2-(methylideneamino)acetonitrile
SMILES C=NCC#N

Specifications

Melting Point 129°C
Color White
Quantity 25 g
Formula Weight 68.08
Physical Form Crystalline Powder
Chemical Name or Material (Methyleneamino)acetonitrile
RTECSNumber MC1925000
TSCA Yes