missing translation for 'onlineSavingsMsg'
Learn More

Methylene Green, Spectrum™ Chemical

$229.25 - $964.75

Chemical Identifiers

CAS 2679-01-8
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611803
View Documents
Spectrum Chemical Mfg Cor
ME1505GM
5 g Amber Glass Bottle
Each for $229.25
Only null left
 
18611802
View Documents
Spectrum Chemical Mfg Cor
ME15025GM
25 g Amber Glass Bottle
Each for $474.04
Only null left
 
18611801
View Documents
Spectrum Chemical Mfg Cor
ME150100GM
100 g Amber Glass Bottle
Each for $964.75
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2679-01-8

Specifications

2679-01-8
Reagent
Dark Brown
Methylene Green
1
Amber Glass Bottle
5 g
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.