missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Solution II, 0.05 mL (1 drop) = 0.1 mg/L Sulfide, Ricca Chemical

Catalog No. R49100001A
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R49100001A 1 L Natural Poly Bottle
49104 120 mL Natural Poly Bottle
491016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R49100001A Supplier Ricca Chemical Company Supplier No. R49100001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Methylene Blue Solution II
Name Note 0.05mL (1 drop) = 0.1mg/L Sulfide
CAS 7220-79-3, 7732-18-5
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L