missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Loeffler Formulation, Loeffler's Alkaline Methylene Blue, Ricca Chemical

For staining and differentiation of acid fast organisms in smears

$102.41 - $102.41

Specifications

Chemical Name or Material METHYLENE BLUE
CAS 7732-18-5,64-17-5,7220-79-3,1310-58-3
CAS Min % 79.0,18.91,0.24,0.01
CAS Max % 82.0,19.68,0.25,0.01
Grade Laboratory
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
4880-16
View Documents
Ricca Chemical Company
488016
500 mL Natural Poly Bottle
Each for $102.41
Only null left
 
Description

Description

Specifications

Specifications

METHYLENE BLUE
79.0,18.91,0.24,0.01
Laboratory
Blue
7732-18-5,64-17-5,7220-79-3,1310-58-3
82.0,19.68,0.25,0.01
Natural Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'