missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue hydrate, 96+%, high purity biological stain

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol

$164.10 - $164.10

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9 hydrate, C.I. 52015 hydrate
PubChem CID 16211647
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC229801000
SDS View Documents
Thermo Scientific Chemicals
229801000
100 g Glass bottle
Each for $164.10
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

122965-43-9
319.85
CXKWCBBOMKCUKX-UHFFFAOYSA-M
16211647
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
C16H18ClN3S
MFCD00150006
Basic Blue 9 hydrate, C.I. 52015 hydrate
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate

Specifications

100.0°C to 110.0°C
96.0 to 103.0% (on dried substance)
C16H18ClN3S
Basic Blue 9 hydrate, C.I. 52015 hydrate
CXKWCBBOMKCUKX-UHFFFAOYSA-M
[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
319.85
8.0% to 22.0% (105°C)
≥96%
Green
Crystalline Powder
122965-43-9
96+%
MFCD00150006
Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol
Authentic
[7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
16211647
319.85
Glass bottle
100 g
Methylene Blue hydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

RTECSNumber : SP5740000

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only