missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, 1% (w/v) in Denatured Alcohol, Ricca Chemical

$176.75 - $216.47

Specifications

Chemical Name or Material Methylene Blue
CAS 64-17-5,67-56-1,7220-79-3
Percent Purity 1% W/V
Grade Laboratory
Packaging Natural Poly Bottle
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
4864-16
View Documents
Ricca Chemical Company
486416
500 mL Natural Poly Bottle
Each for $176.75
Only null left
 
486432
View Documents
Ricca Chemical Company
486432
1 L Natural Poly Bottle
Each for $216.47
Only null left
 
Description

Description

Specifications

Specifications

Methylene Blue
1% W/V
Natural Poly Bottle
64-17-5,67-56-1,7220-79-3
Laboratory
Blue
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'