missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, 1% w/v aq. soln.

CAS: 122965-43-9 | C16H18ClN3S

$61.26 - $106.08

Specifications

Chemical Name or Material Methylene Blue
CAS 122965-43-9,7732-18-5
Assay Percent Range 1% w/v aqueous solution
Molecular Formula C16H18ClN3S
MDL Number MFCD00012111
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42771AE
View Documents
Thermo Scientific Chemicals
042771AE
100 mL
Each for $61.26
Only null left
 
AA42771AP
View Documents
Thermo Scientific Chemicals
042771AP
500 mL
Each for $106.08
Only null left
 
Description

Description

Methylene Blue, 1% w/v aqueous solution is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It is also used in the sulfide analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Methylene Blue
1% w/v aqueous solution
MFCD00012111
319.86
23 hPa (17mm Hg) at 20°C
122965-43-9,7732-18-5
C16H18ClN3S
Miscible with ethanol,chloroform,glacial acetic acid and glycerol. Slightly miscible with pyridine. Immiscible with ethyl ether,xylene,oleic acid,ethanol and acetone.
3, 7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate: 1.0%; Water: 99%
Liquid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only