missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue, 1% in Ethanol, Certified, LabChem™

Supplier:  LabChem LC169202

 View more versions of this product

Catalog No. LC169202


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Passes Test
-115°C
78°C
67-63-0,67-56-1,61-73-4
Soluble in water
319.85
Poly Bottle
Blue
13°C
Methylene Blue
1% in Ethanol
89.1,4.95,4.95,1
C16H18N3SCl
Passes Test
Certified
Carbon monoxide; Carbon dioxide; May release flammable gases
1 L
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions